Assay Details
Target Gene Details
Entrez Gene ID: | 5354 |
Gene Name: | proteolipid protein 1 |
Gene Aliases: |
GPM6C, HLD1, MMPL, PLP, PLP/DM20, PMD, SPG2 |
Location: |
Chr.X:103776506-103792619 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLP1 | NM_000533.4 | 7 | 1099 | NP_000524.3 |
| NM_001128834.2 | 8 | 1181 | NP_001122306.1 | |
| NM_001305004.1 | 7 | 874 | NP_001291933.1 | |
| NM_199478.2 | 7 | 994 | NP_955772.1 | |
| AK128782.1 | 7 | 1022 | ||
| AK292728.1 | ||||
| AK295374.1 | 8 | 952 | ||
| AK295388.1 | 7 | 853 | ||
| AK308966.1 | 6 | 828 | ||
| BC002665.1 | 7 | 937 | AAH02665.1 | |
| BC095452.1 | 7 | 983 | AAH95452.1 | |
| BX445448.2 | 7 | 1000 | ||
| M27110.1 | 7 | 1014 | AAA60117.1 | |
| M54927.1 | 7 | 1017 | AAA59565.1 |
More Information
Set Membership: |
|

Genomic Map