Assay Details
Target Gene Details
Entrez Gene ID: | 55743 |
Gene Name: | checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase |
Gene Aliases: |
RNF116, RNF196 |
Location: |
Chr.12:132840352-132887618 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHFR | NM_001161344.1 | 18 | 2126 | NP_001154816.1 |
| NM_001161345.1 | 18 | 2123 | NP_001154817.1 | |
| NM_001161346.1 | 18 | 2090 | NP_001154818.1 | |
| NM_001161347.1 | 16 | 1850 | NP_001154819.1 | |
| NM_018223.2 | 18 | 2003 | NP_060693.2 | |
| AF170724.1 | 18 | 2132 | AAF91084.1 | |
| AK001658.1 | 18 | 1983 | BAA91817.1 | |
| AK027687.1 | 18 | 2084 | BAB55297.1 | |
| AK054917.1 | 4 | 1031 | ||
| AK090948.1 | 5 | 1099 | ||
| AK097671.1 | 16 | 1849 | ||
| AK302785.1 | ||||
| AK304333.1 | 18 | 2110 | ||
| BC012072.1 | 17 | 2014 | AAH12072.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2661448 | Chr.12:132801861 - 132847994 on Build GRCh38 | Deletion |
|
| dgv318e214 | Chr.12:132801812 - 132847948 on Build GRCh38 | Loss |
|
| nsv561133 | Chr.12:132782284 - 132860654 on Build GRCh38 | Loss |
|
| dgv1589n100 | Chr.12:132841203 - 132967843 on Build GRCh38 | Gain |
|
| nsv455812 | Chr.12:132813412 - 132886923 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map