Assay Details
Target Gene Details
Entrez Gene ID: | 102723809 |
Gene Name: | uncharacterized LOC102723809 |
Gene Aliases: |
- |
Location: |
Chr.14:64514151-64540368 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102723809 | NR_110550.1 | |||
| AI221320.1 | ||||
| BX097562.1 |
Target Gene Details
Entrez Gene ID: | 22890 |
Gene Name: | zinc finger and BTB domain containing 1 |
Gene Aliases: |
ZNF909 |
Location: |
Chr.14:64504574-64533690 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB1 | NM_001123329.1 | 2 | 1688 | NP_001116801.1 |
| NM_014950.2 | 2 | 1688 | NP_055765.2 | |
| XM_005267410.1 | 4 | 1824 | XP_005267467.1 | |
| XM_011536566.2 | 3 | 1561 | XP_011534868.1 | |
| XM_011536568.2 | 4 | 1824 | XP_011534870.1 | |
| XM_017021094.1 | 4 | 2700 | XP_016876583.1 | |
| XM_017021095.1 | 3 | 1616 | XP_016876584.1 | |
| AB023214.1 | 2 | 1559 | ||
| AK291743.1 | 2 | 1466 | ||
| AY598321.1 | 1 | 1297 | AAT06732.1 | |
| BC050719.1 | 4 | 1728 | AAH50719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521823 | Chr.14:64502624 - 64528976 on Build GRCh38 | Loss |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map