Assay Details
Target Gene Details
Entrez Gene ID: | 138009 |
Gene Name: | DDB1 and CUL4 associated factor 4 like 2 |
Gene Aliases: |
WDR21C |
Location: |
Chr.8:87870743-87874068 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCAF4L2 | NM_152418.3 | 1 | 3030 | NP_689631.1 |
| AL833507.1 | 1 | 2978 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831384 | Chr.8:87734490 - 87905020 on Build GRCh38 | Gain |
|
| nsv465734 | Chr.8:86701747 - 87889581 on Build GRCh38 | Gain |
|
| nsv831385 | Chr.8:87795034 - 87953244 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs200647910,rs529590827] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map