Assay Details
Target Gene Details
Entrez Gene ID: | 8669 |
Gene Name: | eukaryotic translation initiation factor 3 subunit J |
Gene Aliases: |
EIF3S1, eIF3-alpha, eIF3-p35 |
Location: |
Chr.15:44537068-44562803 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF3J | NM_001284335.1 | NP_001271264.1 | ||
| NM_001284336.1 | NP_001271265.1 | |||
| NM_003758.3 | NP_003749.2 | |||
| AK002006.1 | ||||
| BU621047.1 | ||||
| U97670.1 | AAC78729.1 |
Target Gene Details
Entrez Gene ID: | 80208 |
Gene Name: | spastic paraplegia 11 (autosomal recessive) |
Gene Aliases: |
ALS5, CMT2X, KIAA1840 |
Location: |
Chr.15:44562696-44663678 on Build GRCh38 |
Assay Gene Location: | Within Exon 41 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPG11 | NM_001160227.1 | 38 | 7331 | NP_001153699.1 |
| NM_025137.3 | 40 | 7670 | NP_079413.3 | |
| XM_006720700.1 | 40 | 7526 | XP_006720763.1 | |
| XM_017022634.1 | 39 | 7562 | XP_016878123.1 | |
| XM_017022636.1 | 29 | 5479 | XP_016878125.1 | |
| AB058743.2 | 40 | 7640 | BAB47469.2 | |
| AK025092.1 | 11 | 1791 | BAB15065.1 | |
| AK057869.1 | 8 | 2010 | ||
| BC024161.1 | 10 | 1756 | ||
| BC067798.1 | 11 | 2032 | AAH67798.1 | |
| BC094704.1 | 11 | 2164 | AAH94704.1 | |
| BC150640.1 | ||||
| BC153879.1 | 40 | 7640 |
More Information
Set Membership: |
|

Genomic Map