Assay Details
Target Gene Details
Entrez Gene ID: | 79665 |
Gene Name: | DEAH-box helicase 40 |
Gene Aliases: |
ARG147, DDX40, PAD |
Location: |
Chr.17:59565525-59608352 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DHX40 | NM_001166301.1 | NP_001159773.1 | ||
| NM_024612.4 | NP_078888.4 | |||
| XM_006722083.2 | XP_006722146.1 | |||
| XM_011525253.2 | XP_011523555.1 | |||
| XM_011525254.2 | XP_011523556.1 | |||
| XM_017025080.1 | XP_016880569.1 | |||
| XM_017025081.1 | XP_016880570.1 | |||
| XM_017025082.1 | XP_016880571.1 | |||
| XM_017025083.1 | XP_016880572.1 | |||
| AF260270.1 | AAF70326.1 | |||
| AF461690.1 | AAN77932.1 | |||
| AK095681.1 | ||||
| AK226048.1 | ||||
| AK299151.1 | ||||
| BC024187.2 | AAH24187.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1067453 | Chr.17:59471819 - 59640766 on Build GRCh38 | Gain |
|
| nsv575805 | Chr.17:59563163 - 59647682 on Build GRCh38 | Gain |
|
| dgv3272n100 | Chr.17:59564145 - 59685102 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map