Assay Details
Target Gene Details
Entrez Gene ID: | 92610 |
Gene Name: | TRAF interacting protein with forkhead associated domain |
Gene Aliases: |
T2BP, T6BP, TIFAA |
Location: |
Chr.4:112275626-112285903 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIFA | NM_052864.2 | 2 | 2853 | NP_443096.1 |
| BM680110.1 | 1 | 238 |
More Information
Set Membership: |
|

Genomic Map