Assay Details
Target Gene Details
Entrez Gene ID: | 64969 |
Gene Name: | mitochondrial ribosomal protein S5 |
Gene Aliases: |
MRP-S5, S5mt |
Location: |
Chr.2:95085391-95122027 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPS5 | NM_001321995.1 | 12 | 1244 | NP_001308924.1 |
| NM_001321996.1 | 12 | 1344 | NP_001308925.1 | |
| NM_001321997.1 | 12 | 1165 | NP_001308926.1 | |
| NM_031902.4 | 12 | 1423 | NP_114108.1 | |
| NR_136152.1 | 11 | 1253 | ||
| NR_136153.1 | 11 | 1332 | ||
| AB049940.1 | 12 | 1304 | BAB40993.1 | |
| AK055390.1 | 2 | 2037 | ||
| AK058160.1 | 11 | 1106 | BAB71695.1 | |
| AK124892.1 | 2 | 3425 | ||
| BC000219.2 | 8 | 738 | AAH00219.2 | |
| BC014172.2 | 12 | 1396 | AAH14172.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160825 | Chr.2:95050220 - 95445574 on Build GRCh38 | Duplication |
|
| dgv4007n100 | Chr.2:94662150 - 95248719 on Build GRCh38 | Gain |
|
| esv3591662 | Chr.2:94986856 - 95146757 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map