Assay Details
Target Gene Details
Entrez Gene ID: | 81579 |
Gene Name: | phospholipase A2 group XIIA |
Gene Aliases: |
GXII, PLA2G12, ROSSY |
Location: |
Chr.4:109709989-109730086 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G12A | NM_030821.4 | 1 | 65 | NP_110448.2 |
| AF332892.1 | AAG50289.1 | |||
| AK075389.1 | 1 | 45 | BAC11589.1 | |
| AK313081.1 | 1 | 49 | ||
| AY359024.1 | AAQ89383.1 | |||
| BC017218.2 | AAH17218.1 | |||
| BI667755.1 | 1 | 70 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv4463 | Chr.4:109726575 - 109771969 on Build GRCh38 | Deletion |
|
| nsv595142 | Chr.4:109688325 - 109735574 on Build GRCh38 | Loss |
|
| esv3569652 | Chr.4:109727450 - 109747513 on Build GRCh38 | Loss |
|
| nsv950375 | Chr.4:109729645 - 109730544 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map