Assay Details
Target Gene Details
Entrez Gene ID: | 146198 |
Gene Name: | ZFP90 zinc finger protein |
Gene Aliases: |
FIK, NK10, ZNF756, zfp-90 |
Location: |
Chr.16:68533484-68576072 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZFP90 | NM_001305203.1 | 5 | 1371 | NP_001292132.1 |
| NM_001305204.1 | NP_001292133.1 | |||
| NM_001305206.1 | 5 | 1426 | NP_001292135.1 | |
| NM_001305207.1 | 5 | 1423 | NP_001292136.1 | |
| NM_001305208.1 | 5 | 1416 | NP_001292137.1 | |
| NM_133458.3 | 5 | 1361 | NP_597715.2 | |
| NR_130976.1 | 5 | 1469 | ||
| NR_130977.1 | 5 | 1419 | ||
| NR_130978.1 | 5 | 1477 | ||
| XM_005255804.2 | 5 | 1301 | XP_005255861.1 | |
| XM_017022952.1 | 4 | 1408 | XP_016878441.1 | |
| AB075834.1 | 1 | 565 | BAB85540.1 | |
| AK056596.1 | 1 | 154 | BAB71228.1 | |
| AK074332.1 | 5 | 1385 | ||
| AK122847.1 | 1 | 1796 | ||
| AK126167.1 | BAC86467.1 | |||
| AK304152.1 | 5 | 1263 | ||
| AK316251.1 | 5 | 1288 | ||
| AW135652.1 | ||||
| BC033165.1 | 1 | 58 | ||
| BC140886.1 | 5 | 1361 | ||
| BC140887.1 | 5 | 1361 | ||
| KC249978.1 |
More Information
Set Membership: |
|

Genomic Map