Assay Details
Target Gene Details
Entrez Gene ID: | 65005 |
Gene Name: | mitochondrial ribosomal protein L9 |
Gene Aliases: |
L9mt |
Location: |
Chr.1:151759643-151763916 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL9 | NM_001300733.1 | NP_001287662.1 | ||
| NM_031420.3 | NP_113608.1 | |||
| NR_125331.1 | ||||
| AB049636.1 | BAB40841.1 | |||
| AK026363.1 | ||||
| AK300668.1 | 4 | 1064 | ||
| AK315459.1 | ||||
| BC004517.2 | AAH04517.1 | |||
| BG437768.1 | ||||
| CF127136.1 |
More Information
Set Membership: |
|

Genomic Map