Assay Details
Target Gene Details
Entrez Gene ID: | 164 |
Gene Name: | adaptor related protein complex 1 gamma 1 subunit |
Gene Aliases: |
ADTG, CLAPG1 |
Location: |
Chr.16:71729000-71809073 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1G1 | NM_001030007.1 | 24 | 5903 | NP_001025178.1 |
| NM_001128.5 | 23 | 5894 | NP_001119.3 | |
| AK025020.1 | 1 | 1594 | ||
| BC003414.1 | 1 | 820 | AAH03414.1 |
More Information
Set Membership: |
|

Genomic Map