Assay Details
Target Gene Details
Entrez Gene ID: | 201501 |
Gene Name: | zinc finger and BTB domain containing 7C |
Gene Aliases: |
APM-1, APM1, ZBTB36, ZNF857C |
Location: |
Chr.18:48026886-48412639 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBTB7C | NM_001039360.2 | 3 | 2829 | NP_001034449.1 |
| NM_001318841.1 | 5 | 3010 | NP_001305770.1 | |
| XM_005258229.4 | 4 | 2754 | XP_005258286.1 | |
| XM_011525861.2 | 7 | 3234 | XP_011524163.1 | |
| XM_011525863.2 | 6 | 3351 | XP_011524165.1 | |
| XM_011525864.2 | 6 | 3106 | XP_011524166.1 | |
| XM_011525865.2 | 6 | 3347 | XP_011524167.1 | |
| XM_011525866.2 | 5 | 3092 | XP_011524168.1 | |
| XM_011525867.2 | 5 | 2881 | XP_011524169.1 | |
| XM_011525869.2 | 6 | 3356 | XP_011524171.1 | |
| XM_011525870.2 | 5 | 3052 | XP_011524172.1 | |
| XM_011525871.2 | 3 | 3567 | XP_011524173.1 | |
| XM_017025605.1 | 7 | 5029 | XP_016881094.1 | |
| XM_017025606.1 | 7 | 3483 | XP_016881095.1 | |
| XM_017025607.1 | 7 | 3238 | XP_016881096.1 | |
| XM_017025608.1 | 6 | 4906 | XP_016881097.1 | |
| XM_017025609.1 | 5 | 3223 | XP_016881098.1 | |
| AK095226.1 | 1 | 566 | ||
| AM988836.1 | 3 | 2817 | ||
| AM988837.1 | 3 | 2962 | ||
| AM988838.1 | 4 | 3012 | ||
| AM988839.1 | 5 | 3010 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv274948 | Chr.18:48025919 - 48031491 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map