Assay Details
Target Gene Details
Entrez Gene ID: | 392 |
Gene Name: | Rho GTPase activating protein 1 |
Gene Aliases: |
CDC42GAP, RHOGAP, RHOGAP1, p50rhoGAP |
Location: |
Chr.11:46677075-46700665 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP1 | NM_004308.3 | 5 | 546 | NP_004299.1 |
| XM_011520095.1 | 5 | 983 | XP_011518397.1 | |
| XM_017017720.1 | XP_016873209.1 | |||
| AK298562.1 | 5 | 493 | ||
| BC018118.1 | 5 | 451 | AAH18118.1 | |
| U02570.1 | 5 | 453 | AAA16142.1 | |
| Z23024.1 | 5 | 480 | CAA80560.1 |
More Information
Set Membership: |
|

Genomic Map