Assay Details
Target Gene Details
Entrez Gene ID: | 246329 |
Gene Name: | SH3 and cysteine rich domain 3 |
Gene Aliases: |
NAM |
Location: |
Chr.12:57243453-57251193 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAC3 | NM_001286256.1 | 11 | 1455 | NP_001273185.1 |
| NM_001286257.1 | 9 | 1089 | NP_001273186.1 | |
| NM_145064.2 | 12 | 1522 | NP_659501.1 | |
| NR_104422.1 | 8 | 1022 | ||
| XM_011538126.2 | 12 | 1610 | XP_011536428.1 | |
| AK057013.1 | 12 | 1515 | BAB71343.1 | |
| AK311513.1 | 8 | 1015 | ||
| BC008069.2 | 11 | 1435 | AAH08069.1 | |
| BU738507.1 | 1 | 147 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
| nsv952189 | Chr.12:57127118 - 57245617 on Build GRCh38 | Deletion |
|
| nsv519293 | Chr.12:57189889 - 57243810 on Build GRCh38 | Loss |
|
| dgv96n21 | Chr.12:57139199 - 57243810 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map