Assay Details
Target Gene Details
Entrez Gene ID: | 8634 |
Gene Name: | RNA 3'-terminal phosphate cyclase |
Gene Aliases: |
RPC, RTC1, RTCD1 |
Location: |
Chr.1:100266158-100292769 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTCA | NM_001130841.1 | NP_001124313.1 | ||
| NM_003729.3 | NP_003720.1 | |||
| XM_005271298.4 | XP_005271355.1 | |||
| XM_005271299.4 | XP_005271356.1 | |||
| XM_017002669.1 | XP_016858158.1 | |||
| AK292366.1 | ||||
| AK299793.1 | ||||
| AK307445.1 | ||||
| AK309394.1 | ||||
| AU310497.1 | ||||
| BC012604.1 | AAH12604.1 | |||
| DA361597.1 | ||||
| Y11651.1 | CAA72364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524345 | Chr.1:100267299 - 100279242 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map