Assay Details
Target Gene Details
Entrez Gene ID: | 240 |
Gene Name: | arachidonate 5-lipoxygenase |
Gene Aliases: |
5-LO, 5-LOX, 5LPG, LOG5 |
Location: |
Chr.10:45374166-45446121 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALOX5 | NM_000698.4 | 14 | 2289 | NP_000689.1 |
| NM_001256153.2 | 14 | 2193 | NP_001243082.1 | |
| NM_001256154.2 | 13 | 2118 | NP_001243083.1 | |
| NM_001320861.1 | 14 | 2202 | NP_001307790.1 | |
| NM_001320862.1 | 14 | 2327 | NP_001307791.1 | |
| XM_017016012.1 | 13 | 2164 | XP_016871501.1 | |
| AW403395.1 | 1 | 328 | ||
| BC130332.1 | 14 | 2242 | ||
| BC132677.1 | 14 | 2242 | ||
| BC143985.1 | 13 | 2071 | ||
| BQ003253.1 | 1 | 295 | ||
| HM592258.1 | ||||
| HM592259.1 | ||||
| HM592260.1 | ||||
| HM592261.1 | ||||
| J03571.1 | 14 | 2209 | AAA65450.1 | |
| J03600.1 | 14 | 2219 | AAA36183.1 |
Target Gene Details
Entrez Gene ID: | 102724323 |
Gene Name: | uncharacterized LOC102724323 |
Gene Aliases: |
- |
Location: |
Chr.10:45444568-45453121 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC102724323 | NR_120674.1 | |||
| AI671881.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1043536 | Chr.10:45420493 - 45580787 on Build GRCh38 | Gain |
|
| esv3891797 | Chr.10:45008026 - 45674466 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map