Assay Details
Target Gene Details
Entrez Gene ID: | 57214 |
Gene Name: | cell migration inducing hyaluronan binding protein |
Gene Aliases: |
CCSP1, HYBID, KIAA1199, TMEM2L |
Location: |
Chr.15:80779343-80951662 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CEMIP | NM_001293298.1 | 30 | 5316 | NP_001280227.1 |
| NM_001293304.1 | 30 | 5256 | NP_001280233.1 | |
| NM_018689.2 | 29 | 5157 | NP_061159.1 | |
| AB033025.1 | 21 | 3825 | BAA86513.1 | |
| AB103330.1 | 29 | 5129 | BAD02451.1 | |
| AK000465.1 | ||||
| AK026540.1 | 14 | 2706 | ||
| AK027058.1 | 1 | 820 | ||
| AL049389.1 | 1 | 355 | ||
| AL359061.1 | 17 | 3200 | CAB94391.1 | |
| AY007811.1 | 10 | 2288 | AAG41059.1 | |
| AY581148.1 | 30 | 5288 | AAU06219.1 | |
| AY581149.1 | 30 | 5228 | AAU06220.1 | |
| AY585237.1 | 29 | 5129 | AAU06221.1 |
Target Gene Details
Entrez Gene ID: | 23184 |
Gene Name: | mesoderm development candidate 2 |
Gene Aliases: |
BOCA, MESD |
Location: |
Chr.15:80947326-80989878 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MESDC2 | NR_126327.1 | |||
| NR_126328.1 | ||||
| AK301416.1 | ||||
| BC009210.2 | AAH09210.1 | |||
| BC012746.2 | AAH12746.1 |
More Information
Set Membership: |
|

Genomic Map