Assay Details
Target Gene Details
Entrez Gene ID: | 53836 |
Gene Name: | G protein-coupled receptor 87 |
Gene Aliases: |
FKSG78, GPR95, KPG_002 |
Location: |
Chr.3:151294086-151316848 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR87 | NM_023915.3 | 3 | 523 | NP_076404.3 |
| AB031548.1 | 3 | 455 | BAD83583.1 | |
| AF237763.1 | 3 | 411 | AAK01858.1 | |
| AF345566.1 | 1 | 201 | AAK29069.1 | |
| AF411114.1 | 2 | 162 | AAL26485.1 | |
| AK027784.1 | 3 | 523 | BAB55366.1 | |
| BC009540.1 | AAH09540.2 | |||
| BC112941.1 | 3 | 432 | AAI12942.1 | |
| BC117565.1 | 3 | 432 | ||
| BC132888.1 | 3 | 489 | ||
| BC132890.1 | 3 | 489 | ||
| BP226924.1 | ||||
| EU883576.1 | 2 | 162 |
Target Gene Details
Entrez Gene ID: | 116931 |
Gene Name: | mediator complex subunit 12 like |
Gene Aliases: |
NOPAR, TNRC11L, TRALP, TRALPUSH |
Location: |
Chr.3:151085665-151436677 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED12L | NM_053002.5 | NP_443728.3 | ||
| XM_006713487.3 | XP_006713550.1 | |||
| XM_011512390.2 | XP_011510692.1 | |||
| XM_011512394.2 | XP_011510696.1 | |||
| XM_011512399.2 | XP_011510701.1 | |||
| XM_017005676.1 | XP_016861165.1 | |||
| XM_017005677.1 | XP_016861166.1 | |||
| XM_017005678.1 | XP_016861167.1 | |||
| XM_017005679.1 | XP_016861168.1 | |||
| XM_017005680.1 | XP_016861169.1 | |||
| AB046855.1 | BAB13461.1 | |||
| AF399708.1 | AAO38813.1 | |||
| AK128165.1 | ||||
| BC144559.1 | ||||
| BX108326.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv592045 | Chr.3:151052827 - 151377305 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map