Assay Details
Target Gene Details
Entrez Gene ID: | 53358 |
Gene Name: | SHC adaptor protein 3 |
Gene Aliases: |
N-Shc, NSHC, RAI, SHCC |
Location: |
Chr.9:89005771-89182308 on Build GRCh38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHC3 | NM_016848.5 | 12 | 2116 | NP_058544.3 |
| XM_011518785.2 | 12 | 5658 | XP_011517087.1 | |
| XM_011518786.2 | 13 | 1507 | XP_011517088.1 | |
| BC026314.1 | 12 | 2117 | AAH26314.1 | |
| BX641139.1 | 8 | 4549 | ||
| D84361.1 | 12 | 1989 | BAA12323.1 | |
| KU176125.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3621014 | Chr.9:88877891 - 89641453 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map