Assay Details
Target Gene Details
Entrez Gene ID: | 10644 |
Gene Name: | insulin like growth factor 2 mRNA binding protein 2 |
Gene Aliases: |
IMP-2, IMP2, VICKZ2 |
Location: |
Chr.3:185643739-185825051 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF2BP2 | NM_001007225.1 | NP_001007226.1 | ||
| NM_001291869.1 | NP_001278798.1 | |||
| NM_001291872.1 | NP_001278801.1 | |||
| NM_001291873.1 | NP_001278802.1 | |||
| NM_001291874.1 | NP_001278803.1 | |||
| NM_001291875.1 | NP_001278804.1 | |||
| NM_006548.4 | NP_006539.3 | |||
| XM_017005557.1 | XP_016861046.1 | |||
| XM_017005558.1 | XP_016861047.1 | |||
| XM_017005559.1 | XP_016861048.1 | |||
| XM_017005560.1 | XP_016861049.1 | |||
| XM_017005561.1 | XP_016861050.1 | |||
| XM_017005562.1 | XP_016861051.1 | |||
| XM_017005563.1 | XP_016861052.1 | |||
| XM_017005564.1 | XP_016861053.1 | |||
| AF117107.1 | AAD09827.1 | |||
| AK290664.1 | ||||
| AK309611.1 | ||||
| BC021290.2 | AAH21290.1 | |||
| EU408701.1 | ||||
| EU408702.1 | ||||
| EU408703.1 | ||||
| EU408704.1 |
Target Gene Details
Entrez Gene ID: | 646600 |
Gene Name: | IGF2BP2 antisense RNA 1 |
Gene Aliases: |
C3orf65 |
Location: |
Chr.3:185713252-185729787 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF2BP2-AS1 | NR_126326.1 | |||
| AK057462.1 | 2 | 1526 | BAB71499.1 |
More Information
Set Membership: |
|

Genomic Map