Assay Details
Target Gene Details
Entrez Gene ID: | 29966 |
Gene Name: | striatin 3 |
Gene Aliases: |
PPP2R6B, SG2NA |
Location: |
Chr.14:30893799-31026528 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 17 - Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STRN3 | NM_001083893.1 | NP_001077362.1 | ||
| NM_014574.3 | NP_055389.3 | |||
| XM_005267569.3 | XP_005267626.1 | |||
| XM_005267570.3 | XP_005267627.1 | |||
| AF243424.1 | AAF81201.1 | |||
| AK290229.1 | ||||
| AK314041.1 | ||||
| BC126221.1 | ||||
| BC132673.1 | ||||
| U17989.1 | AAB81551.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv564169 | Chr.14:30349881 - 31072288 on Build GRCh38 | Gain |
|
| nsv832762 | Chr.14:30769428 - 30944832 on Build GRCh38 | Loss |
|
| dgv3605n54 | Chr.14:30889171 - 30944286 on Build GRCh38 | Loss |
|
| esv33189 | Chr.14:30331017 - 31442539 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map