Assay Details
Target Gene Details
Entrez Gene ID: | 2307 |
Gene Name: | forkhead box S1 |
Gene Aliases: |
FKHL18, FREAC10 |
Location: |
Chr.20:31844300-31845617 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FOXS1 | NM_004118.3 | 1 | 410 | NP_004109.1 |
| AF042831.1 | AAC15420.1 | |||
| BC013408.2 | 1 | 410 | AAH13408.1 | |
| BT009768.1 | 1 | 335 | AAP88770.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv4300n100 | Chr.20:31247234 - 32033468 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map