Assay Details
Target Gene Details
Entrez Gene ID: | 6273 |
Gene Name: | S100 calcium binding protein A2 |
Gene Aliases: |
CAN19, S100L |
Location: |
Chr.1:153561108-153565830 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| S100A2 | NM_005978.3 | 3 | 588 | NP_005969.1 |
| AF086003.1 | 2 | 243 | ||
| BC002829.2 | 3 | 588 | ||
| BC105787.1 | 3 | 292 | AAI05788.1 | |
| M87068.1 | 3 | 290 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831603 | Chr.1:153433366 - 153603667 on Build GRCh38 | Gain |
|
| esv3587591 | Chr.1:153528630 - 153570193 on Build GRCh38 | Gain |
|
| esv3587584 | Chr.1:153466126 - 153598991 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map