Assay Details
Target Gene Details
| Entrez Gene ID: | 51649 | 
| Gene Name: | mitochondrial ribosomal protein S23 | 
| Gene Aliases: | CGI-138, HSPC329, MRP-S23 | 
| Location: | Chr.17:57838926-57850072 on Build GRCh38 | 
| Assay Gene Location: | Overlaps - Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MRPS23 | NM_016070.3 | NP_057154.2 | ||
| BX479815.1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Non-exonic  non-DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top