Assay Details
Target Gene Details
Entrez Gene ID: | 57826 |
Gene Name: | RAP2C, member of RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.X:132203024-132219480 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2C | NM_001271186.1 | 6 | 3591 | NP_001258115.1 |
| NM_001271187.1 | 6 | 2926 | NP_001258116.1 | |
| NM_021183.4 | 4 | 3519 | NP_067006.3 | |
| NR_073150.1 | 7 | 2967 | ||
| XM_006724775.2 | 5 | 3249 | XP_006724838.1 | |
| XM_011531376.1 | 6 | 3659 | XP_011529678.1 | |
| AK124801.1 | 3 | 2829 | ||
| AL049685.2 | 3 | 2751 | CAB41256.1 | |
| AL832461.1 | 3 | 3070 | ||
| AY298955.1 | 4 | 3519 | AAP55684.1 | |
| BC003403.1 | 3 | 2805 | AAH03403.1 | |
| BX647223.1 | 4 | 2808 | ||
| DB535605.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map