Assay Details
Target Gene Details
Entrez Gene ID: | 85236 |
Gene Name: | histone cluster 1, H2bk |
Gene Aliases: |
H2B/S, H2BFAiii, H2BFT, H2BK |
Location: |
Chr.6:27138293-27146858 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H2BK | NM_080593.2 | NP_542160.1 | ||
| AJ223352.1 | CAA11276.1 | |||
| BC000893.1 | AAH00893.1 | |||
| BC051872.1 | AAH51872.1 | |||
| BC064959.1 | AAH64959.1 | |||
| BC108737.1 | AAI08738.1 | |||
| DA873786.1 |
Target Gene Details
Entrez Gene ID: | 8294 |
Gene Name: | histone cluster 1, H4i |
Gene Aliases: |
H4/m, H4FM, H4M |
Location: |
Chr.6:27139309-27139678 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HIST1H4I | NM_003495.2 | NP_003486.1 | ||
| BC066250.1 | AAH66250.1 | |||
| BC067496.1 | AAH67496.1 | |||
| BC067497.1 | AAH67497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1029124 | Chr.6:26857689 - 27141995 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map