Assay Details
Target Gene Details
Entrez Gene ID: | 3479 |
Gene Name: | insulin like growth factor 1 |
Gene Aliases: |
IGF-I, IGFI, MGF |
Location: |
Chr.12:102395867-102481839 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IGF1 | NM_000618.4 | 4 | 5423 | NP_000609.1 |
| NM_001111283.2 | 5 | 5472 | NP_001104753.1 | |
| NM_001111284.1 | 4 | 5261 | NP_001104754.1 | |
| XM_017019262.1 | 5 | 5282 | XP_016874751.1 | |
| XM_017019263.1 | 4 | 5225 | XP_016874752.1 | |
| X57025.1 | 4 | 5325 | CAA40342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832498 | Chr.12:102334798 - 102519791 on Build GRCh38 | Gain |
|
| nsv560034 | Chr.12:102397524 - 102400754 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map