Assay Details
Target Gene Details
Entrez Gene ID: | 29922 |
Gene Name: | NME/NM23 family member 7 |
Gene Aliases: |
CFAP67, MN23H7, NDK 7, NDK7, nm23-H7 |
Location: |
Chr.1:169132385-169367963 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NME7 | NM_013330.4 | NP_037462.1 | ||
| NM_197972.2 | NP_932076.1 | |||
| NR_104229.1 | ||||
| XM_005245106.4 | XP_005245163.1 | |||
| XM_017001100.1 | XP_016856589.1 | |||
| AF153191.2 | AAD34622.1 | |||
| AK094513.1 | ||||
| AK290701.1 | ||||
| AK301917.1 | ||||
| BC006983.1 | AAH06983.1 | |||
| BG204249.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv22143 | Chr.1:169090390 - 169652030 on Build GRCh38 | Gain+Loss |
|
| nsv831858 | Chr.1:169053145 - 169209830 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map