Assay Details
Target Gene Details
Entrez Gene ID: | 1116 |
Gene Name: | chitinase 3 like 1 |
Gene Aliases: |
ASRT7, CGP-39, GP-39, GP39, HC-gp39, HCGP-3P, YKL-40, YKL40, YYL-40, hCGP-39 |
Location: |
Chr.1:203178931-203186794 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CHI3L1 | NM_001276.2 | 10 | 1663 | NP_001267.2 |
| AB209459.1 | 1 | 6073 | BAD92696.1 | |
| AK095458.1 | 2 | 1746 | ||
| AK130142.1 | 10 | 1573 | ||
| AK130200.1 | 10 | 1573 | ||
| AK225266.1 | 10 | 1572 | ||
| AK225584.1 | 10 | 1572 | ||
| BC008568.1 | 10 | 1609 | AAH08568.1 | |
| BC038354.1 | 10 | 1550 | AAH38354.1 | |
| BC039132.1 | 10 | 1559 | AAH39132.1 | |
| BX648308.1 | 3 | 3345 | ||
| M80927.1 | 10 | 1563 | AAA16074.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952639 | Chr.1:203164873 - 203186672 on Build GRCh38 | Deletion |
|
| nsv832370 | Chr.1:202997191 - 203196105 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map