Assay Details
Target Gene Details
Entrez Gene ID: | 2444 |
Gene Name: | fyn related Src family tyrosine kinase |
Gene Aliases: |
GTK, PTK5, RAK |
Location: |
Chr.6:115941031-116100739 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FRK | NM_002031.2 | 5 | 1331 | NP_002022.1 |
| XM_005266880.4 | 6 | 1403 | XP_005266937.1 | |
| XM_005266881.2 | 6 | 1320 | XP_005266938.1 | |
| XM_005266882.4 | 7 | 1632 | XP_005266939.1 | |
| XM_011535653.2 | 7 | 1635 | XP_011533955.1 | |
| XM_011535654.2 | 6 | 1417 | XP_011533956.1 | |
| XM_011535655.2 | 6 | 1414 | XP_011533957.1 | |
| XM_011535656.2 | 5 | 788 | XP_011533958.1 | |
| XM_017010645.1 | 6 | 1521 | XP_016866134.1 | |
| AK302264.1 | 4 | 506 | ||
| AK313314.1 | 5 | 1411 | ||
| BC012916.1 | 5 | 1176 | AAH12916.1 | |
| U00803.1 | 5 | 1331 | AAA18284.1 | |
| U22322.1 | 5 | 1293 | AAC50116.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3610466 | Chr.6:115941729 - 115968686 on Build GRCh38 | Loss |
|
| nsv1029998 | Chr.6:115763322 - 116057888 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map