Assay Details
Target Gene Details
Entrez Gene ID: | 7113 |
Gene Name: | transmembrane protease, serine 2 |
Gene Aliases: |
PP9284, PRSS10 |
Location: |
Chr.21:41464551-41508158 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMPRSS2 | NM_001135099.1 | 14 | 2851 | NP_001128571.1 |
| NM_005656.3 | 14 | 2813 | NP_005647.3 | |
| XM_005261043.2 | 13 | 2693 | XP_005261100.1 | |
| XM_011529733.1 | 12 | 2512 | XP_011528035.1 | |
| AA904080.1 | ||||
| AF318374.1 | 1 | 1906 | AAL55881.1 | |
| AF329454.1 | 14 | 2806 | AAK53559.1 | |
| AK123948.1 | 1 | 2147 | ||
| BC051839.1 | 14 | 2784 | AAH51839.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv459261 | Chr.21:41458763 - 41494180 on Build GRCh38 | Loss |
|
| nsv834102 | Chr.21:41286267 - 41494006 on Build GRCh38 | Loss |
|
| esv3647063 | Chr.21:41455441 - 41527402 on Build GRCh38 | Gain |
|
| esv3647064 | Chr.21:41456009 - 41569886 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map