Assay Details
Target Gene Details
Entrez Gene ID: | 10526 |
Gene Name: | importin 8 |
Gene Aliases: |
RANBP8 |
Location: |
Chr.12:30628981-30695995 on Build GRCh38 |
Assay Gene Location: | Within Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IPO8 | NM_001190995.1 | 21 | 4299 | NP_001177924.1 |
| NM_006390.3 | 25 | 4937 | NP_006381.2 | |
| XM_017018691.1 | 25 | 4834 | XP_016874180.1 | |
| XM_017018692.1 | 24 | 4494 | XP_016874181.1 | |
| AB209602.1 | 11 | 3954 | BAD92839.1 | |
| AK000201.1 | 1 | 525 | ||
| AK025607.1 | 1 | 2368 | ||
| AK130090.1 | 5 | 2116 | ||
| BG574795.1 | 1 | 261 |
More Information
Additional Information:
For this assay, SNP(s) [rs74450923,rs77388133] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map