Assay Details
Target Gene Details
Entrez Gene ID: | 29978 |
Gene Name: | ubiquilin 2 |
Gene Aliases: |
ALS15, CHAP1, DSK2, HRIHFB2157, N4BP4, PLIC2 |
Location: |
Chr.X:56563593-56567010 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UBQLN2 | NM_013444.3 | 1 | 962 | NP_038472.2 |
| AB015344.1 | BAA34801.1 | |||
| AF189009.1 | 1 | 916 | AAF17237.1 | |
| AF293385.1 | 1 | 681 | AAG02474.1 | |
| AK297252.1 | 1 | 928 | ||
| AK302885.1 | 1 | 928 | ||
| AL442081.1 | 1 | 575 | CAC09446.1 | |
| BC069237.1 | 2 | 833 | AAH69237.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6916 | Chr.X:56527462 - 56572321 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map