Assay Details
Target Gene Details
Entrez Gene ID: | 2919 |
Gene Name: | C-X-C motif chemokine ligand 1 |
Gene Aliases: |
FSP, GRO1, GROa, MGSA, MGSA-a, NAP-3, SCYB1 |
Location: |
Chr.4:73869392-73871302 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL1 | NM_001511.3 | 4 | 670 | NP_001502.1 |
| NR_046035.1 | 4 | 693 | ||
| BC011976.1 | 4 | 658 | AAH11976.1 | |
| BM554448.1 | 4 | 683 | ||
| BM996663.1 | ||||
| BQ000742.1 | 1 | 450 | ||
| J03561.1 | 4 | 644 | AAA35933.1 | |
| X12510.1 | 4 | 608 | CAA31027.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1001725 | Chr.4:73488333 - 73995207 on Build GRCh38 | Gain |
|
| nsv525980 | Chr.4:73865809 - 74556070 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map