Assay Details
Target Gene Details
Entrez Gene ID: | 80310 |
Gene Name: | platelet derived growth factor D |
Gene Aliases: |
IEGF, MSTP036, SCDGF-B, SCDGFB |
Location: |
Chr.11:103907186-104164299 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDGFD | NM_025208.4 | 7 | 3645 | NP_079484.1 |
| NM_033135.3 | 7 | 3627 | NP_149126.1 | |
| AF113216.1 | 7 | 3379 | AAG39287.1 | |
| AI962915.1 | 1 | 353 | ||
| AY027517.1 | 7 | 3365 | AAK20081.1 | |
| AY027518.1 | 7 | 3347 | AAK20082.1 | |
| BC030645.2 | 7 | 3707 | AAH30645.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2750977 | Chr.11:103211829 - 104054062 on Build GRCh38 | Gain |
|
| esv2760322 | Chr.11:103889357 - 104503413 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map