Assay Details
Target Gene Details
Entrez Gene ID: | 1130 |
Gene Name: | lysosomal trafficking regulator |
Gene Aliases: |
CHS, CHS1 |
Location: |
Chr.1:235661031-235883708 on Build GRCh38 |
Assay Gene Location: | Within Exon 60 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LYST | NM_000081.3 | 53 | 11725 | NP_000072.2 |
| NM_001301365.1 | 53 | 12162 | NP_001288294.1 | |
| XM_011544031.1 | 54 | 11887 | XP_011542333.1 | |
| XM_011544032.1 | 54 | 12290 | XP_011542334.1 | |
| XM_011544033.2 | 54 | 12037 | XP_011542335.1 | |
| XM_011544034.1 | 53 | 11749 | XP_011542336.1 | |
| XM_011544036.2 | 50 | 9541 | XP_011542338.1 | |
| XM_017000150.1 | 53 | 11656 | XP_016855639.1 | |
| BX649113.1 | 22 | 6535 | ||
| U67615.1 | 53 | 11732 | AAB41309.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1011288 | Chr.1:235376656 - 235666302 on Build GRCh38 | Gain |
|
| nsv832936 | Chr.1:235644298 - 235809947 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map