Assay Details
Target Gene Details
Entrez Gene ID: | 9057 |
Gene Name: | solute carrier family 7 member 6 |
Gene Aliases: |
LAT-2, LAT3, y+LAT-2 |
Location: |
Chr.16:68263830-68301823 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A6 | NM_001076785.2 | NP_001070253.1 | ||
| NM_003983.5 | NP_003974.3 | |||
| XM_011523433.1 | XP_011521735.1 | |||
| XM_011523434.1 | XP_011521736.1 | |||
| XM_011523438.1 | XP_011521740.1 | |||
| XM_011523439.1 | XP_011521741.1 | |||
| XM_017023851.1 | XP_016879340.1 | |||
| CR749291.1 | CAH18146.1 | |||
| CR749475.1 | CAH18305.1 | |||
| D87432.1 | ||||
| R48943.1 |
Target Gene Details
Entrez Gene ID: | 84138 |
Gene Name: | solute carrier family 7 member 6 opposite strand |
Gene Aliases: |
- |
Location: |
Chr.16:68300615-68310965 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A6OS | NM_032178.2 | NP_115554.2 | ||
| AK023353.1 | BAB14538.1 | |||
| AK091674.1 | ||||
| AK225712.1 | ||||
| AY078070.1 | AAL79958.1 | |||
| BC013778.1 | AAH13778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510691 | Chr.16:68266527 - 68325431 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs544214685] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map