Assay Details
Target Gene Details
Entrez Gene ID: | 205564 |
Gene Name: | SUMO1/sentrin specific peptidase 5 |
Gene Aliases: |
- |
Location: |
Chr.3:196867856-196934714 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SENP5 | NM_001308045.1 | 2 | 1063 | NP_001294974.1 |
| NM_152699.4 | 2 | 1063 | NP_689912.2 | |
| XM_005269310.2 | 2 | 1063 | XP_005269367.1 | |
| XM_005269312.2 | 2 | 1063 | XP_005269369.1 | |
| XM_011512544.2 | 3 | 1108 | XP_011510846.1 | |
| XM_011512545.2 | 2 | 1063 | XP_011510847.1 | |
| XM_017005870.1 | 3 | 1063 | XP_016861359.1 | |
| AK124389.1 | ||||
| AK302305.1 | 2 | 1063 | ||
| AK313764.1 | 2 | 1036 | ||
| BC008589.1 | 1 | 160 | AAH08589.1 | |
| BC030705.2 | 2 | 1012 | AAH30705.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3599293 | Chr.3:196759100 - 197444213 on Build GRCh38 | Gain |
|
| dgv5036n100 | Chr.3:196792338 - 196926122 on Build GRCh38 | Gain |
|
| nsv1001045 | Chr.3:196703082 - 196900384 on Build GRCh38 | Gain |
|
| nsv829829 | Chr.3:196747858 - 196887060 on Build GRCh38 | Loss |
|
| nsv1003135 | Chr.3:196621850 - 197012082 on Build GRCh38 | Gain |
|
| esv2763775 | Chr.3:196727069 - 197015419 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map