Assay Details
Target Gene Details
Entrez Gene ID: | 9572 |
Gene Name: | nuclear receptor subfamily 1 group D member 1 |
Gene Aliases: |
EAR1, THRA1, THRAL, ear-1, hRev |
Location: |
Chr.17:40092784-40100725 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NR1D1 | NM_021724.4 | 5 | 1298 | NP_068370.1 |
| AK300155.1 | ||||
| AK315270.1 | ||||
| AY429554.1 | ||||
| BC047875.1 | 5 | 1122 | ||
| BC056148.1 | 5 | 1189 | AAH56148.1 | |
| HQ692861.1 | 5 | 667 | ||
| M24898.1 | 5 | 1294 | AAA52335.1 | |
| X72631.1 | 5 | 869 | CAB53540.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833444 | Chr.17:40086570 - 40281028 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map