Assay Details
Target Gene Details
Entrez Gene ID: | 4801 |
Gene Name: | nuclear transcription factor Y subunit beta |
Gene Aliases: |
CBF-A, CBF-B, HAP3, NF-YB |
Location: |
Chr.12:104117080-104138262 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYB | NM_006166.3 | 8 | 1839 | NP_006157.1 |
| XM_005268908.4 | 9 | 1903 | XP_005268965.1 | |
| XM_005268909.4 | 9 | 1886 | XP_005268966.1 | |
| XM_005268910.4 | 9 | 2558 | XP_005268967.1 | |
| CD678671.1 | 1 | 378 |
More Information
Additional Information:
For this assay, SNP(s) [rs78519510] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map