Assay Details
Target Gene Details
Entrez Gene ID: | 9271 |
Gene Name: | piwi like RNA-mediated gene silencing 1 |
Gene Aliases: |
CT80.1, HIWI, MIWI, PIWI |
Location: |
Chr.12:130337887-130412708 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIWIL1 | NM_001190971.1 | NP_001177900.1 | ||
| NM_004764.4 | NP_004755.2 | |||
| XM_011539002.2 | XP_011537304.1 | |||
| XM_011539003.2 | XP_011537305.1 | |||
| XM_011539004.2 | XP_011537306.1 | |||
| XM_011539005.1 | XP_011537307.1 | |||
| XM_017020229.1 | XP_016875718.1 | |||
| AB274731.1 | ||||
| AF104260.2 | AAC97371.2 | |||
| AF264004.1 | AAK92281.1 | |||
| AF387507.1 | AAK69348.1 | |||
| AK093133.1 | BAC04068.1 | |||
| BC028581.2 | AAH28581.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3582027 | Chr.12:130227822 - 130722407 on Build GRCh38 | Gain |
|
| esv34208 | Chr.12:129528244 - 130603882 on Build GRCh38 | Gain |
|
| nsv560735 | Chr.12:129710094 - 130478603 on Build GRCh38 | Gain |
|
| dgv2977n54 | Chr.12:130222462 - 130829669 on Build GRCh38 | Gain |
|
| esv2759921 | Chr.12:129461919 - 130631031 on Build GRCh38 | Gain+Loss |
|
| nsv1044735 | Chr.12:130295681 - 130367666 on Build GRCh38 | Gain |
|
| nsv455765 | Chr.12:130341310 - 130399802 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map