Assay Details
Target Gene Details
Entrez Gene ID: | 8634 |
Gene Name: | RNA 3'-terminal phosphate cyclase |
Gene Aliases: |
RPC, RTC1, RTCD1 |
Location: |
Chr.1:100266158-100292769 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RTCA | NM_001130841.1 | 8 | 917 | NP_001124313.1 |
| NM_003729.3 | 7 | 878 | NP_003720.1 | |
| XM_005271298.4 | 7 | 875 | XP_005271355.1 | |
| XM_005271299.4 | 7 | 864 | XP_005271356.1 | |
| XM_017002669.1 | 7 | 621 | XP_016858158.1 | |
| AK292366.1 | 7 | 827 | ||
| AK307445.1 | 5 | 614 | ||
| AK309394.1 | ||||
| BC012604.1 | 8 | 831 | AAH12604.1 | |
| Y11651.1 | 7 | 830 | CAA72364.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv524345 | Chr.1:100267299 - 100279242 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map