Assay Details
Target Gene Details
Entrez Gene ID: | 84064 |
Gene Name: | haloacid dehalogenase like hydrolase domain containing 2 |
Gene Aliases: |
3110052N05Rik, HEL-S-301 |
Location: |
Chr.18:47107402-47150520 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HDHD2 | NM_001318765.1 | 7 | 1876 | NP_001305694.1 |
| NM_032124.4 | 7 | 1819 | NP_115500.1 | |
| XM_006722557.2 | 8 | 1911 | XP_006722620.1 | |
| XM_011526227.2 | 8 | 1895 | XP_011524529.1 | |
| XM_011526228.2 | 8 | 1893 | XP_011524530.1 | |
| XM_017026039.1 | 8 | 1932 | XP_016881528.1 | |
| AA206788.1 | 1 | 403 | ||
| AK054590.1 | 7 | 1856 | BAB70768.1 | |
| AL136681.1 | 7 | 1810 | CAB66616.1 | |
| BC033031.1 | 7 | 1855 | AAH33031.1 | |
| BC038100.2 | 7 | 1819 | AAH38100.1 | |
| BX537623.1 | 1 | 3081 | ||
| FJ224322.1 | 7 | 1810 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3642451 | Chr.18:47089642 - 47219390 on Build GRCh38 | Gain |
|
| nsv576819 | Chr.18:47075878 - 47151256 on Build GRCh38 | Loss |
|
| nsv9629 | Chr.18:46684730 - 47231140 on Build GRCh38 | Gain+Loss |
|
| nsv833640 | Chr.18:47025905 - 47188501 on Build GRCh38 | Loss |
|
| esv3642452 | Chr.18:47093035 - 47161369 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map