Assay Details
Target Gene Details
Entrez Gene ID: | 286187 |
Gene Name: | protein phosphatase 1 regulatory subunit 42 |
Gene Aliases: |
LRRC67, TLLR, dtr |
Location: |
Chr.8:66988131-67028551 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPP1R42 | NM_001013626.2 | NP_001013648.1 | ||
| BC055413.1 | AAH55413.1 |
More Information
Set Membership: |
|

Genomic Map