Assay Details
Target Gene Details
Entrez Gene ID: | 346653 |
Gene Name: | family with sequence similarity 71 member F2 |
Gene Aliases: |
FAM137B |
Location: |
Chr.7:128671628-128702262 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71F2 | NM_001012454.5 | NP_001012457.3 | ||
| NM_001128926.3 | NP_001122398.1 | |||
| NM_001290254.1 | NP_001277183.1 | |||
| NM_001290255.1 | NP_001277184.1 | |||
| XM_011516128.2 | XP_011514430.1 | |||
| XM_011516129.2 | XP_011514431.1 | |||
| XM_011516130.2 | XP_011514432.1 | |||
| XM_011516131.2 | XP_011514433.1 | |||
| XM_011516132.2 | XP_011514434.1 | |||
| XM_011516134.2 | XP_011514436.1 | |||
| XM_011516135.2 | XP_011514437.1 | |||
| XM_011516136.2 | XP_011514438.1 | |||
| XM_017012147.1 | XP_016867636.1 | |||
| XM_017012149.1 | XP_016867638.1 | |||
| BC047310.1 | AAH47310.2 | |||
| BC066973.1 | AAH66973.2 | |||
| BC105729.1 | ||||
| BC105730.1 | ||||
| BC105731.1 | ||||
| BI828876.1 | ||||
| HY039605.1 | ||||
| HY046252.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2670312 | Chr.7:128499519 - 128999613 on Build GRCh38 | Deletion |
|
| esv2735137 | Chr.7:128499440 - 128999664 on Build GRCh38 | Deletion |
|

Genomic Map