Assay Details
Target Gene Details
Entrez Gene ID: | 3688 |
Gene Name: | integrin subunit beta 1 |
Gene Aliases: |
CD29, FNRB, GPIIA, MDF2, MSK12, VLA-BETA, VLAB |
Location: |
Chr.10:32900318-32958365 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ITGB1 | NM_002211.3 | 10 | 1428 | NP_002202.2 |
| NM_033668.2 | 9 | 1207 | NP_391988.1 | |
| NM_133376.2 | 10 | 1343 | NP_596867.1 | |
| AK291697.1 | 10 | 1280 | ||
| BC020057.1 | 10 | 1272 | AAH20057.1 | |
| BC113901.1 | AAI13902.1 | |||
| BM791591.1 | ||||
| BX537407.1 | 10 | 1282 | CAD97649.1 | |
| CX868374.1 | 6 | 685 | ||
| CX871886.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831827 | Chr.10:32817738 - 32985904 on Build GRCh38 | Loss |
|
| nsv550383 | Chr.10:32898536 - 32960205 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map