Assay Details
Target Gene Details
Entrez Gene ID: | 84225 |
Gene Name: | zinc finger MYND-type containing 15 |
Gene Aliases: |
SPGF14 |
Location: |
Chr.17:4740015-4746119 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZMYND15 | NM_001136046.2 | 2 | 288 | NP_001129518.1 |
| NM_001267822.1 | 2 | 288 | NP_001254751.1 | |
| NM_032265.2 | 2 | 288 | NP_115641.1 | |
| XM_017025218.1 | 2 | 279 | XP_016880707.1 | |
| AK302181.1 | 2 | 279 | ||
| AK310469.1 | 1 | 741 | ||
| AL136893.1 | 1 | 261 | CAB66827.1 | |
| DB453494.2 | 2 | 288 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv574273 | Chr.17:4585589 - 5460332 on Build GRCh38 | Gain |
|
| nsv833346 | Chr.17:4646256 - 4860871 on Build GRCh38 | Loss |
|
| nsv1952 | Chr.17:4726245 - 4748889 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map