Assay Details
Target Gene Details
Entrez Gene ID: | 80018 |
Gene Name: | N(alpha)-acetyltransferase 25, NatB auxiliary subunit |
Gene Aliases: |
C12orf30, MDM20, NAP1 |
Location: |
Chr.12:112026689-112109022 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA25 | NM_024953.3 | 19 | 2349 | NP_079229.2 |
| XM_005253938.1 | 18 | 2195 | XP_005253995.1 | |
| XM_006719606.2 | 19 | 3009 | XP_006719669.1 | |
| XM_017019977.1 | 17 | 2065 | XP_016875466.1 | |
| AB054990.1 | 19 | 2291 | BAC80174.1 | |
| AK023151.1 | 16 | 1994 | BAB14432.1 | |
| AK292485.1 | 19 | 2322 | ||
| AK307494.1 | 20 | 2498 | ||
| AK309772.1 | 19 | 2304 | ||
| AL832394.1 | 3 | 417 | ||
| BC034357.1 | 19 | 2303 | ||
| BC113585.1 | 19 | 2349 | ||
| BC113587.1 | 19 | 2349 | ||
| BX641140.1 | 19 | 2304 | CAE46062.1 | |
| BX647450.1 | 19 | 2302 | ||
| BX647992.1 | 17 | 2079 |
More Information
Set Membership: |
|

Genomic Map