Assay Details
Target Gene Details
Entrez Gene ID: | 586 |
Gene Name: | branched chain amino acid transaminase 1 |
Gene Aliases: |
BCATC, BCT1, ECA39, MECA39, PNAS121, PP18 |
Location: |
Chr.12:24810024-24949459 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCAT1 | NM_001178091.1 | 10 | 6768 | NP_001171562.1 |
| NM_001178092.1 | 9 | 6696 | NP_001171563.1 | |
| NM_001178093.1 | 11 | 6475 | NP_001171564.1 | |
| NM_001178094.1 | 11 | 6772 | NP_001171565.1 | |
| NM_005504.6 | 11 | 6879 | NP_005495.2 | |
| XM_017019768.1 | 10 | 6487 | XP_016875257.1 | |
| AK025615.1 | 1 | 1845 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557794 | Chr.12:24811449 - 24817774 on Build GRCh38 | Gain |
|
| esv2422312 | Chr.12:24366458 - 25068752 on Build GRCh38 | Duplication |
|
| dgv2412n54 | Chr.12:24738241 - 25054719 on Build GRCh38 | Gain |
|
| nsv557775 | Chr.12:24539449 - 25204746 on Build GRCh38 | Gain |
|
| nsv819279 | Chr.12:24812523 - 24812963 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map